Anti-BMP4 Rabbit Polyclonal Antibody

Supplier: Prosci

49-648
10079-128EA 582.9 USD
10079-128
Anti-BMP4 Rabbit Polyclonal Antibody
Antibodies
Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Through molecular cloning, 7 BMP cDNAs, designated BMP1 through BMP7, have been recovered. BMP2 through BMP7, are members of the transforming growth factor-beta superfamily of regulatory molecules. Transcript variants utilizing alternative promoters and alternative non-coding first exons have been described in the literature.

BMP4 antibody can be used in ELISA starting at 1:5000 - 1:25000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence.

Protein G Column

PBS, 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: BMP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR