Anti-UCHL1 Chicken Polyclonal Antibody

Supplier: ProSci
49-639
10079-116EA 749.47 USD
10079-116
Anti-UCHL1 Chicken Polyclonal Antibody
Antibodies
UCHL1 is a member of family of proteins that hydrolyze small C-terminal adducts of ubiquitin to generate the ubiquitin monomer. The expression of UCHL1 is highly specific to neurons and to cell of the diffuse neuroendocrine system and their tumors. UCHL1 exhibits both and hydrolase and a ligase activity. The S18Y variant of UCHL1, that has reduced ligase activity, has been linked to susceptibility for Parkinson’s disease.

UCHL1 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

PBS. No preservative added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR