Anti-LY86 Rabbit Polyclonal Antibody

Supplier: Prosci

49-615
10079-096EA 647.66 USD
10079-096
Anti-LY86 Rabbit Polyclonal Antibody
Antibodies
LY86 antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LY86
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR