Anti-AP3S2 Chicken Polyclonal Antibody

Supplier: ProSci
49-596
10079-072EA 707.44 USD
10079-072
Anti-AP3S2 Chicken Polyclonal Antibody
Antibodies
AP3S2 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS. No preservative added.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: AP3S2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR