Anti-SETDB1 Rabbit Polyclonal Antibody

Supplier: ProSci
49-593
10076-458EA 674.53 USD
10076-458
Anti-SETDB1 Rabbit Polyclonal Antibody
Antibodies
The SET domain is a highly conserved, approximately 150-amino acid motif implicated in the modulation of chromatin structure. It was originally identified as part of a larger conserved region present in the Drosophila trithorax protein and was subsequently identified in the Drosophila Su(var)3-9 and 'Enhancer of zeste' proteins, from which the acronym SET is derived. Studies have suggested that the SET domain may be a signature of proteins that modulate transcriptionally active or repressed chromatin states through chromatin remodeling activities. ESET functions as a histone methyltransferase by methylation of Lys-9 of histone H3. H3 Lys-9 methylation represents a specific tag for epigenetic transcriptional repression by recruiting HP1 proteins to methylated histones. ESET shows a nuclear localization and is associated with non-pericentromeric regions of chromatin, and is excluded from nucleoli and islands of condensed chromatin. Although ESET is a widely expressed protein, it is highly expressed in the testis.

SETDB1 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SETDB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR