Anti-MAP4K4 Rabbit Polyclonal Antibody

Supplier: Prosci

49-584
10076-440EA 624.31 USD
10076-440
Anti-MAP4K4 Rabbit Polyclonal Antibody
Antibodies
MAP4K4 is a MSN protein kinase. The protein has a structure similar to other GCK (Group I) kinases and contains two consensus SH3 binding motifs, which have been shown to interact with the SH3 domains of the SH2/SH3 adapter protein Nck. NIK can be activated by tumor necrosis factor (TNF)-alpha. Activated NIK has been shown to activate c-Jun N-terminal kinase (JNK) and to bind and activate the plasma membrane Na-H exchanger NHE1. In NIK(-/-) mice, mesodermal and endodermal cells of the primitive streak fail to migrate to their correct location, and embryos die between embryonic day (E) 9.5 and E10.5. At least three protein isoforms have been reported.

MAP4K4 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

Purified

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MAP4K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR