Anti-FANCA Rabbit Polyclonal Antibody

Supplier: Prosci

49-573
10076-432EA 582.9 USD
10076-432
Anti-FANCA Rabbit Polyclonal Antibody
Antibodies
FANCA (also called Protein FACA or Fanconi anemia group A protein) is involved in DNA repair, perhaps specifically with post-replication repair or a cell cycle checkpoint function. FANCA may also be implicated in interstrand DNA cross-link repair and in the maintenance of normal chromosome stability. The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, and FANCL. The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group A. Alternative splicing results in multiple transcript variants encoding different isoforms. Variant 1 (isoform a) encodes the longest transcript. Variant 2 (isoform b) contains an alternate exon, which results in an early stop codon, compared to variant 1. Isoform b has a shorter C-terminus when compared to isoform a. Mutations in this gene are the most common cause of Fanconi anemia.

FANCA antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 20 μg/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FANCA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR