Anti-PDCD4 Rabbit Polyclonal Antibody

Supplier: ProSci
49-547
10076-408EA 711.98 USD
10076-408
Anti-PDCD4 Rabbit Polyclonal Antibody
Antibodies
Programmed cell death 4 (Pdcd4) is a novel tumor supressor. Pdcd4 directly inhibits the helicase activity of eukaryotic translation initiation factor 4A (eIF4A), a component of the translation initiation complex. Pdcd4 also suppresses the transactivation of activator protein-1 (AP-1)-responsive promoters by c-Jun. Pdcd4 contains two Akt phosphorylation sites, one at Ser67 and the other at Ser457. The phosphorylation of Pdcd4 by Akt causes nuclear translocation of Pdcd4 and a significant decrease in the ability of Pdcd4 to interfere with the transactivation of AP-1-responsive promoters by c-Jun.

PDCD4 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PDCD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Xenopus, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR