Anti-CDKN2A Rabbit Polyclonal Antibody

Supplier: ProSci
49-544
10076-402EA 727.25 USD
10076-402
Anti-CDKN2A Rabbit Polyclonal Antibody
Antibodies
Found to be homozygously deleted at high frequency in cell lines derived from tumors of lung, breast, brain, bone, skin, bladder, kidney, ovary, and lymphocyte. Mutations are also associated with pancreatic adenocarcinoma.

CDKN2A antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

Delipidation and Defibrination

0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CDKN2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR