Anti-ANGPT1 Rabbit Polyclonal Antibody

Supplier: Prosci

49-525
10076-384EA 582.9 USD
10076-384
Anti-ANGPT1 Rabbit Polyclonal Antibody
Antibodies
Angiopoeitin-1 (Ang-1) and Angiopoietin-2 (Ang-2) have importance in the development of the endothelium by regulating tyrosine phosphorylation of the membrane receptor Tie-2/Tek. Ang-1 binding to Tie-2/Tek causes phosphorylation of the receptor. Ang-2 competes for this binding, and thus blocks receptor phosphorylation. Ang-1 has a potential fibrinogen-like domain at the carboxyl terminus and coiled-coil regions in the amino terminus. Ang-1 is prominently expressed in the myocardium of atrium and ventricle, mesenchymal and smooth muscle cells surrounding most blood vessels, and lung. In the adult, Ang-1 is also expressed in the heart and liver.

Angiopoietin-1 antibody can be used in ELISA starting at 1 ?g/mL, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

Sterile Filtration

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ANGPT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR