Anti-SFRP1 Rabbit Polyclonal Antibody

Supplier: ProSci
49-520
10076-378EA 722.46 USD
10076-378
Anti-SFRP1 Rabbit Polyclonal Antibody
Antibodies
SFRP1 (also known as FRP, FRP1, SARP2, Secreted Apoptosis-related Protein 2, Secreted Frizzled-related Protein and Secreted frizzled-related protein 1) is a member of the SFRP family that contains a cysteine-rich domain homologous to the putative Wnt-binding site of Frizzled proteins. SFRPs act as soluble modulators of Wnt signaling. SFRP1 and SFRP5 may be involved in determining the polarity of photoreceptor cells in the retina. SFRP1 is expressed in several human tissues, with the highest levels in heart.

SFRP1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SFRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR