Anti-S1PR5 Rabbit Polyclonal Antibody

Supplier: Prosci

49-514
10079-064EA 582.9 USD
10079-064
Anti-S1PR5 Rabbit Polyclonal Antibody
Antibodies
Edg8 is a Lysophospholipid/Lysosphingolipid Receptor activated by sphingosine-1-phosphate. Two differently expressed splice variants, 2.4 kb and 5.4 kb, are produced by alternative splicing.

EDG8 antibody can be used in ELISA starting at 1:10000 - 1:40000, Western Blot starting at 1:200 - 1:2000, immunohistochemistry starting at 2.5 ?g/mL, immunofluorescence, and immunoprecipitation.

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: S1PR5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR