Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NOXO1 Rabbit Polyclonal Antibody

Supplier: Prosci
49-506
The enzymes NADPH oxidase (NOX) and dual oxidase (DUOX) generate ROS in a regulated manner, producing reactive oxygen in various cells and tissues in response to growth factors, cytokines and calcium signals. The oxidase consists of the catalytic subunit gp91phox (otherwise known as NOX2), together with the regulatory subunits p22phox, p47phox, p40phox, p67phox and the small GTPase RAC. The enzyme activity of gp91phox is regulated by the assembly of these regulatory subunits with gp91phox to form an active complex. In 1999, the first of the NOX homologues of gp91phox was described as NOX1. The enzyme was cloned from a colon epithelial cell complementary DNA library. When expressed in cells, NOX1 generated low amounts of ROS, but high-level ROS production by NOX1 was subsequently achieved by co-expression with novel regulatory subunits (described later). Subsequently, NOX3 and NOX4 were cloned, and the latter was shown to generate high levels of ROS when expressed in cells. NOX organizer 1 (NOXO1) is a homologue of p47phox and has an almost identical domain organization, except that it lacks the auto-inhibitory region. NOX activator 1 (NOXA1) is a homologue of p67phox and similarly shares the same domain organization, except that it lacks one of the two SH3 domains that are present in p67phox. Co-transfection of NOX1, NOXO1 and NOXA1 results in marked ROS generation. Similar to p47phox, NOXO1 binds to p22phox, which is required for NOX1-dependent activity. NOXA1 has a well-conserved activation domain, implying a conserved mechanism for regulating the activity of the target NOX enzyme. Taken from: Lambeth JD. Nat Rev Immunol. 2004 Mar;4:181-9.

NOXO1 antibody can be used in Western Blot starting at 2 - 3 μg/mL, and immunohistochemistry starting at 5 μg/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Store NOXO1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles. Store undiluted.

Type:
Antigen: NOXO1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 µg 49-506 10079-052 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol NOXO1
Antigen Name NADPH oxidase organizer 1
Reactivity Chimpanzee, Human
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Antigen Synonyms P41NOXC, P41NOXB, P41NOX, Nox organizer 1, SNX28, MGC20258, NADPH oxidase organizer 1, P41NOXA, SH3PXD5, regulatory protein P41NOX, NOXO1
Immunogen NOXO1 antibody was raised against amino acids 238 - 252 of NOXO1 (Human).
Form Liquid
Tested Applications Immunoaffinity Chromatography

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty