Anti-TF Rabbit Polyclonal Antibody

Supplier: Prosci

49-489
10079-026EA 582.9 USD
10079-026
Anti-TF Rabbit Polyclonal Antibody
Antibodies
Transferrin (TF) is a glycoprotein with an approximate molecular weight of 76.5 kDa. TF is thought to have been created as a result of an ancient gene duplication event that led to generation of homologous C and N-terminal domains each of which binds 1 ion of ferric iron. The function of transferrin is to transport iron from the intestine, reticuloendothelial system, and liver parenchymal cells to all proliferating cells in the body. In addition to its function in iron transport, transferrin may also have a physiologic role as granulocyte/pollen-binding protein (GPBP) involved in the removal of certain organic matter/allergins from serum.

Serotransferrin antibody can be used in immunohistochemistry starting at 4 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR