Anti-SPANXC Rabbit Polyclonal Antibody

Supplier: Prosci

49-487
10079-022EA 615.27 USD
10079-022
Anti-SPANXC Rabbit Polyclonal Antibody
Antibodies
Human Sperm Proteins Associated with the Nucleus on X-chromosome (SPANX) are relatively low molecular weight cytoplasmic proteins found in testis and sperm. Their expression in other tissues indicates malignancies. Family members are observed as proteins that range from 15 to 20 kDa.

SPANXC antibody can be used in Western Blot, and immunohistochemistry starting at 10 μg/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SPANXC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR