Anti-SKP1 Rabbit Polyclonal Antibody

Supplier: ProSci
49-485
10079-018EA 711.98 USD
10079-018
Anti-SKP1 Rabbit Polyclonal Antibody
Antibodies
SKP1 is also known as S-phase kinase-associated protein 1A, Cyclin A/CDK2-associated protein p19, p19A, p19skp1, RNA polymerase II elongation factor-like protein, Organ of Corti protein 2, OCP-II protein, OCP-2, Transcription elongation factor B, and SIII. SKP1 is an essential component of the SCF (SKP1-CUL1-F-box protein) ubiquitin ligase complex, which mediates the ubiquitination of proteins involved in cell cycle progression, signal transduction and transcription. In the SCF complex, SKP1 serves as an adapter that links the F-box protein to CUL1. SKP1 interacts directly with CUL1 and F-box proteins, such as BTRC and SKP2, in the SCF complex. SKP1 also interacts with FBX29 and the cyclin A/CDK2 complex. SKP1 is part of a SCF-like complex consisting of CUL7, RBX1, SKP1 and FBXW8 and is also a component of a E3 ubiquitin ligase complex containing UBE2D1, SIAH1, CACYBP/SIP, SKP1A, APC and TBL1X.

SKP1 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

Delipidation and Defibrination

Contains 0.01% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SKP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR