Anti-RBX1 Rabbit Polyclonal Antibody

Supplier: Prosci

49-477
10079-008EA 582.9 USD
10079-008
Anti-RBX1 Rabbit Polyclonal Antibody
Antibodies
ROC1 also known as RING-box protein 1, Rbx1, Regulator of cullins 1, RING finger protein 75, and ZYP protein, is a component of the SCF (SKP1-CUL1-F-box protein) and the CBC(VHL) (CUL2-elonging BC-VHL) E3 ubiquitin ligase complexes, which mediate the ubiquitination and subsequent proteasomal degradation of target proteins involved in cell cycle progression, signal transduction and transcription. ROC1 appears to recruit the E2 ubiquitination enzyme through the RING-type zinc finger in a manner similar to CDC34, and brings it into close proximity to the substrate. The RING-type zinc finger domain is essential for ubiquitin ligase activity. ROC1 probably also stimulates CDC34 autoubiquitination and promotes the neddylation of CUL1 and probably CUL2. ROC1 has a cytoplasmic and nuclear localization and is widely expressed in most tissues.

Ring-box 1 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

Delipidation and Defibrination

0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RBX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR