Anti-POLM Rabbit Polyclonal Antibody

Supplier: ProSci
49-467
10078-994EA 722.46 USD
10078-994
Anti-POLM Rabbit Polyclonal Antibody
Antibodies
POLM antibody can be used in ELISA, and immunohistochemistry starting at 20 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: POLM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR