Anti-NF2 Rabbit Polyclonal Antibody

Supplier: Prosci

49-453
10078-974EA 582.9 USD
10078-974
Anti-NF2 Rabbit Polyclonal Antibody
Antibodies
The recently isolated neurofibromatosis 2 tumor suppressor gene encodes a 595 amino acid protein (Merlin). The protein product Merlin, named for its relatedness to the ezrin, radixin and moesin (ERM) family of proteins, is a tumor suppressor whose absence results in the occurrence of multiple tumors of the nervous system, particularly schwannomas and meningiomas. Merlin's similarity to ERMs suggests that it might share functions, acting as a link between cytoskeletal components and the cell membrane. The NF2 protein is highly expressed in human fibroblasts and is detected as a single band of about 75 kDa. This antibody is specific for the phosphorylated form of human NF2 (Merlin).

Neurofibromin 2 antibody can be used in ELISA, and immunohistochemistry starting at 2.5 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NF2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR