Anti-IRAK1 Rabbit Polyclonal Antibody

Supplier: ProSci
49-435
10078-948EA 674.53 USD
10078-948
Anti-IRAK1 Rabbit Polyclonal Antibody
Antibodies
IRAK (Interleukin-1 receptor-associated kinase 1). Nuclear factor kappa B (NF-κB) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-κB mediates the expression of a great variety of genes in response to extracellular stimuli including IL-1, TNFa and LPS. A serine/ threonine protein kinase associated with IL-1 receptor (IRAK) and its homologue mouse pelle-like protein kinase (mPLK) were identified recently (1,2). IRAK is associated with the IL-1 receptor subunits IL-1RI and IL-1RAcP after IL-1 binding and serves as a signaling molecule to mediate IL-1 response. IRAK mediates a signaling cascade leading to NF-κB activation by members in IL-1 family including IL-1 and a novel cytokine IL-18 (also termed IGIF) (1,4).

IRAK1 antibody can be used in immunohistochemistry starting at 10 ?g/mL, immunoprecipitation, and flow cytometry.

IgG Fractionation

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IRAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR