Anti-FBXO9 Rabbit Polyclonal Antibody

Supplier: ProSci
49-428
10078-934EA 734.51 USD
10078-934
Anti-FBXO9 Rabbit Polyclonal Antibody
Antibodies
F-box only protein 9 (also called FBOX9 and Fbp9) is a member of the F-box protein family, which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class. Alternative splicing of this gene generates at least 3 transcript variants diverging at the 5' terminus. Isoform 1 is distinct from isoforms.

FBXO9 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, immunohistochemistry starting at 1:100, immunohistochemistry in frozen tissues starting at 10 - 20 ?g/mL, and flow cytometry.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FBXO9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR