Anti-C5AR1 Mouse Monoclonal Antibody

49-426
10078-932EA 491.23 USD
10078-932
Anti-C5AR1 Mouse Monoclonal Antibody
C5a Anaphylatoxin Receptor mediates the effects of one of the most potent chemotactic agents known, in a wide variety of inflammatory cell types. C5a receptor 1 has been implicated in asthma.

C5AR1 antibody can be used in Western Blot starting at 1 - 2 ?g/mL, immunohistochemistry starting at 10 ?g/mL, immunoprecipitation starting at 2 µg/mg lysate, and immunofluorescence.

Ammonium Sulfate Precipitation

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: 0
Antigen: C5AR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Bovine, Ferret, Human, Mustelid, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR