FANCC (also called Protein FACC or Fanconi Anemia Group C protein) is involved in DNA repair, perhaps specifically with post-replication repair or a cell cycle checkpoint function. FANCC may also be implicated in interstrand DNA cross-link repair and in the maintenance of normal chromosome stability. FANCC belongs to the multi-subunit Fanconi Anemia (FA) complex composed of FANCA, FANCB, FANCC, FANCE, FANCF, FANCG, FANCL/PHF9 and FANCM. FANCC is mainly found within the nucleus although some protein is localized in the cytoplasm. This protein is ubiquitously expressed. Defects in FANCC are a cause of Fanconi anemia (FA). FA is a genetically heterogeneous, autosomal recessive disorder characterized by progressive pancytopaenia, a diverse assortment of congenital malformations, and a predisposition to the development of malignancies. At the cellular level it is associated with hypersensitivity to DNA-damaging agents, chromosomal instability (increased chromosome breakage), and defective DNA repair.
FANCC antibody can be used in ELISA starting at 0.3 ?g/mL, Western Blot, and immunohistochemistry starting at 5 ?g/mL.
0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: FANCC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Chimpanzee, Human