Anti-CCNT1 Rabbit Polyclonal Antibody

Supplier: Prosci

49-406
10078-892EA 615.27 USD
10078-892
Anti-CCNT1 Rabbit Polyclonal Antibody
Antibodies
Cyclin T1, together with the kinase CDK9, is a component of the transcription elongation factor P-TEFb which binds the human immunodeficiency virus type 1 (HIV-1) transactivator Tat. Tat stimulates human HIV-1 viral transcription elongation. This suggests that cyclin T1/cdk9(PITALRE) is one of the HIV-1 required host cellular cofactors generated during T cell activation. Cyclin T1/cdk9(PITALRE) is shown to interact with Tat to restore Tat activation in HeLa nuclear extracts depleted of P-TEFb. P-TEFb facilitates transcription by phosphorylating the carboxy-terminal domain (CTD) of RNA polymerase II. The cdk9(PITALRE) activity and cyclin T1 are essential for activation of transcription when tethered to the heterologous Rev response element RNA via the regulator of expression of virion Rev. Cyclin T1 is an exceptionally large cyclin and is therefore a candidate for interactions with regulatory proteins.

Cyclin T1 antibody can be used in Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation.

Sterile Filtration

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CCNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR