Islet of insulinoma amyloid polypeptide (IAPP) is a 37 amino acids monomeric polypeptide isolated from pancreatic amyloid. It is commonly found in the islet of patients with diabetes mellitus type II and in insulinomas. IAPP selectively inhibits insulin-stimulated glucose utilization and glycogen deposition in muscle, while not affecting adipocyte glucose metabolism.
IAPP antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.
Tissue Culture Supernatant
0.09% sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: IAPP
Clonality: Monoclonal
Clone: [R10/99]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human