Anti-COL3A1 Rabbit Polyclonal Antibody (Biotin)

49-394
10078-878EA 491.23 USD
10078-878
Anti-COL3A1 Rabbit Polyclonal Antibody (Biotin)
COL3A1 antibody can be used in Western Blot starting at 1:2000, and immunohistochemistry starting at 2.5 ?g/mL.

0.125 M sodium borate, 0.075 M NaCl, 0.005 M EDTA;, pH 8.0, 0.01% sodium azide, 10 mg/mL BSA (Immunoglobulin, protease Free).

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: COL3A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR