Anti-ANAPC10 Rabbit Polyclonal Antibody

Supplier: ProSci
49-381
10078-860EA 711.98 USD
10078-860
Anti-ANAPC10 Rabbit Polyclonal Antibody
Antibodies
APC is a ubiquitin ligase which specifically targets mitotic regulatory factors such as Pds 1/Cut 2 and cyclin B. It was found that APC 10/Doc 1 is localized in centrosomes and mitotic spindles throughout mitosis, while it is also localized in kinetochores from prophase to anaphase and in mid body in telophase and cytokinesis. These results strongly support the notion that human APC 10/Doc 1 may be one of the APC core subunits rather than the transiently associated regulatory factor.

ANAPC10 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ANAPC10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR