Anti-ALPI Rabbit Polyclonal Antibody (Biotin)

Supplier: ProSci
49-380
10078-858EA 774.57 USD
10078-858
Anti-ALPI Rabbit Polyclonal Antibody (Biotin)
Antibodies
ALPI antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

0.01 % sodium azide, 0.02 M potassium phosphate, 0.15 M NaCl, dH2O.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ALPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR