Anti-MTOR Rabbit Polyclonal Antibody

Supplier: Prosci

49-374
10078-846EA 615.27 USD
10078-846
Anti-MTOR Rabbit Polyclonal Antibody
Antibodies
Mammalian target of rapamycin (mTOR) is a serine and threonine protein kinase that regulates numerous cellular functions, in particular, the initiation of protein translation. Rapamycin is a natural product macrolide that induces G1 growth arrest in yeast, Drosophila, and mammalian cells. mTOR has a long list of synonyms including FK506 binding protein12 - rapamycin associated protein 1, FK506 binding protein12 - rapamycin associated protein 2, FRAP1, FRAP2, RAFT1, RAPT1 and/or FKBP12-rapamycin associated protein (FRAP). mTOR is one of a family of proteins involved in cell cycle progression, DNA recombination, and DNA damage detection. In rat, mTOR is a 245-kD protein referred to as RAFT1 with significant homology to the Saccharomyces cerevisiae protein TOR1 and has been shown to associate with the immunophilin FKBP12 in a rapamycin-dependent fashion. The FKBP12-rapamycin complex is known to inhibit progression through the G1 cell cycle stage by interfering with mitogenic signaling pathways involved in G1 progression in several cell types, as well as in yeast. The binding of mTOR to FKBP12-rapamycin correlates with the ability of these ligands to inhibit cell cycle progression.

FRAP1 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 2.5 μg/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR