Anti-SLC2A4 Rabbit Polyclonal Antibody

Supplier: Prosci

49-370
10078-842EA 647.66 USD
10078-842
Anti-SLC2A4 Rabbit Polyclonal Antibody
Antibodies
Glucose Transporter 4 (GLUT4/SLC2A4) is a member of a family of proteins that facilitate glucose transport. In response to insulin GLUT4 translocates from low density microsomes to the plasma membrane. Activation of phosphatidylinositol-3-OH kinase is required for this trafficking event but is not sufficient. Insulin resistant subjects with and without diabetes show abnormal accumulation of GLUT4 in a dense membrane compartment in muscle and adipocytes from which insulin is unable to recruit GLUT4 to the cell surface.

SLC2A4 antibody can be used in immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 1:25 - 1:50.

Affinity Chromatography

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SLC2A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Monkey, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR