Anti-GOT1 Sheep Polyclonal Antibody

Supplier: ProSci
49-368
10078-840EA 785.68 USD
10078-840
Anti-GOT1 Sheep Polyclonal Antibody
Antibodies
Glutamic-oxaloacetic transaminase is a pyridoxal phosphate-dependent enzyme which exists in cytoplasmic and inner-membrane mitochondrial forms, GOT1 and GOT2, respectively. GOT plays a role in amino acid metabolism and the urea and tricarboxylic acid cycles. The two enzymes are homodimeric and show close homology.

GOT2 antibody can be used in Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation.

IgG Fractionation

0.015 M potassium phosphate buffer, 0.15 M NaCl, pH 7.2, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GOT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR