Anti-MLF1IP Rabbit Polyclonal Antibody

Supplier: ProSci
49-359
10078-832EA 734.51 USD
10078-832
Anti-MLF1IP Rabbit Polyclonal Antibody
Antibodies
Myeloid leukemia factor-1 (MLF1) Interacting Protein (also known as PBIP1, MLF1IP1, KLIP1 or KSHV latent nuclear antigen interacting protein 1) is a novel polo-like kinase 1 (Plk1) substrate. Plk1 phosphorylation of MLF1IP induces ubiquitination and degradation of MLF1IP prior to the metaphase/anaphase transition. Several Plk1-dependent phosphorylation sites have been identified on MLF1IP by mass spectrometry. Mutations of these sites stabilize MLF1IP and inhibit mitotic progression. Subsequent in vitro and in vivo MLF1IP phosphorylation and stability assays have revealed that phosphorylation of Thr78 is critical for triggering Plk1-dependent MLF1IP degradation. Expression of a non-degradable Thr78Ala mutant was sufficient to induce a mitotic block. Timely phosphorylation of MLF1IP on Thr78 by Plk1 is critical for eliminating the MLF1IP-imposed mitotic block prior to anaphase onset. MLF1IP is speculated to be a novel tumor suppressor, whose function is required for proper sister-chromatid separation. Loss of MLF1IP function may result in improper segregation of chromosomes and genomic instability, thus promoting tumorigenesis.

MLF1immunoprecipitation antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MLF1IP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Dog, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR