Anti-RELB Rabbit Polyclonal Antibody

49-358
10078-830EA 540.97 USD
10078-830
Anti-RELB Rabbit Polyclonal Antibody
RELB antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RELB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR