Anti-SARM1 Mouse Monoclonal Antibody [clone: [Sarmy-1]]

Supplier: ProSci
49-353
10078-822EA 698.16 USD
10078-822
Anti-SARM1 Mouse Monoclonal Antibody [clone: [Sarmy-1]]
Antibodies
Toll-like receptors (TLRs) play an essential role in the detection and elimination of invading microbes. They are type-1 transmembrane receptors, containing extracellular leucine rich repeats and an intracellular Toll/interleukin-1 receptor (TIR) domain. Upon stimulation, these receptors interact with specific TIR domain-containing adaptor proteins. Five such adaptors have been discovered to date, MyD88, Mal (MyD88 adaptor-like)/TIRAP (TIR domain-containing adaptor protein), Trif (TIR-domain-containing adaptor inducing interferon-beta), TRAM (Trif-related adaptor molecule) and SARM (SAM and ARM-containing protein). Different TLRs use different combinations of these adaptors, leading to the activation of common and unique pathways involved in the elimination of the invading microbe.

SARM1 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

Purified

PBS, 10% glycerol, 0.02% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SARM1
Clonality: Monoclonal
Clone: [Sarmy-1]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR