Anti-OPTN Rabbit Polyclonal Antibody

Supplier: ProSci
49-349
10078-820EA 684.46 USD
10078-820
Anti-OPTN Rabbit Polyclonal Antibody
Antibodies
Tumor Necrosis Factor Alpha-inducible Cellular Protein Containing Leucine Zipper Domains; Huntingtin Interacting Protein L; Transcrption Factor IIIA-interacting Protein (FIP2) (OPTN) Rabbit anti-Human Polyclonal Antibody

OPTN antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

TBS, pH 7.4, 50% glycerol, 0.5 mg/mL BSA, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: OPTN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gorilla, Human, Monkey, Orangutan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR