Anti-TNFAIP3 Interacting Protein 2 Rabbit Polyclonal Antibody

Supplier: Prosci

49-334
10078-796EA 582.9 USD
10078-796
Anti-TNFAIP3 Interacting Protein 2 Rabbit Polyclonal Antibody
Antibodies
ABIN-2 binds to the COOH-terminal zinc finger domain of A20. NF-κB activation induced by TNF and interleukin-1 is inhibited by overexpression of ABIN-2. The latter also inhibits NF-κB activation induced by overexpression of receptor-interacting protein or TNF receptor-associated factor 2. A-20 binding Inhibitor of Nuclear Factor-kappaB (NF-κB)Activation (ABIN-2) binds to A-20. ABIN-2 interferes with NF-κB activation upstream of the IKK complex and might contribute to NF-κB inhibitory function of A-20.

TNimmunoprecipitation2 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 2.5 ?g/mL.

Protein G Column

PBS, 0.05% BSA, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR