Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFRSF19 Rabbit Polyclonal Antibody

Supplier: Prosci
49-330
TAJ/TROY is a membrane protein of 423 amino acids with characteristic cysteine-rich motifs in the extracellular domain and a tumor necrosis factor receptor-associated factor (TRAF) 2 binding sequence in the cytoplasmic domain. The extracellular domain of TROY has significant homology with that of EDAR (another newly identified receptor belonging to TNF receptor family) that specifies hair follicle fate. The cytoplasmic domain of TAJ is different than other TNF receptor family members. Although its cytoplasmic domain does not contain death domain, TAJ/TROY induced cell death is probably mediated through a caspase-independent pathway. TAJ-induced JNK activation was not blocked by dominant-negative inhibitors of TRAF2, TRAF5, or ASK1, which have been previously implicated in JNK activation via TNFR1 and CD40. However, coimmunoprecipitation assays revealed that TAJ is capable of binding a number of different TRAF family members, and it is possible that TAJ-induced JNKactivation is mediated by an as yet untested TRAF homolog, such as TRAF6. TNFRSF19, also known as TROY, TRADE or TAJ, is a member of the tumor necrosis factor receptor superfamily. It has the typical TNFR superfamily extracellular ligand-binding domain but lacks an intracellular death domain. TAJ has been shown to activate the c-Jun N-terminal kinase pathway and mediate Caspase-independent cell death Eby et al. (2000). TAJ has been shown by ISH to be expressed in the epithelium of many embryonic mouse tissues may play a role in embryonic development Kojima et al. (2000).

TNFRSF19 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 μg/mL.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TNFRSF19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 µg 49-330 10076-364 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol TNFRSF19
Antigen Name Tumor Necrosis Factor Receptor Superfamily Member 19
Reactivity Mouse, Human
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
Isotype IgG
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms AL023044, Troy, TAJ, TAJ-ALPHA, TRADE, AW123854
Immunogen TNFRSF19 antibody was raised against amino acids 29 - 44 of TNFRSF19 (Human).
Form Liquid

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty