Anti-OLFM4 Rabbit Polyclonal Antibody

Supplier: Prosci

49-306
10076-316EA 582.9 USD
10076-316
Anti-OLFM4 Rabbit Polyclonal Antibody
Antibodies
Olfactomedin 4 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

PBS, 0.05% BSA, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: OLFM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR