Anti-IL20RA Rabbit Polyclonal Antibody

Supplier: ProSci
49-295
10076-294EA 674.53 USD
10076-294
Anti-IL20RA Rabbit Polyclonal Antibody
Antibodies
Principal Names: Interleukin 20 receptor alpha(IL20RA); IL-20R1; ZcytoR7; ZCYTOR7; Cytokine receptor Class II member 8(CRF2-8); Official Gene Symbol: IL20RA Gene ID: 53832 (Human) Gene Map Locus: 6q22.33-q23.1 (Human) IL20RA,is a subunit of IL20R, a member of Cytokine Class II receptor super family. It consists of a central transmembrane domain and functionally induces IL20 and IL26 signaling in epithelial cells and keratinocytes. It associates with IL-20RB and forms a functional receptor complexfor IL20, IL19, and IL24. It also associates with IL-10RB and forms a unique heterodimeric functional receptor complex for IL-26. Upon ligand binding, it induces signaling through activation of STAT3 and STAT1. IL20RA is specifically expressed in skin suggesting a prominent role in growth and proliferation of keratinocytes.

IL-20A Receptor antibody can be used in immunohistochemistry starting at 15 ?g/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IL20RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR