Anti-HTR3A Rabbit Polyclonal Antibody

Supplier: ProSci
49-292
10076-288EA 674.53 USD
10076-288
Anti-HTR3A Rabbit Polyclonal Antibody
Antibodies
Serotonin (5-hydroxytryptamine, 5-HT), originally discovered as a serum factor plays important roles in regulating diverse biological processes in central and peripheral nervous systems, cardiovascular systems, and gastrointestinal systems. The 5-HT3 receptors are distinguished from other 5-HT receptors by their structure and function. 5-HT3 receptors span the membrane only four times and are ligand-gated, cation-selective ion channels that mediate neuronal excitation and exocytotic release of neurotransmitters within the peripheral and central nervous systems. Functional studies done on isolated tissues have revealed heterogeneity of this receptor. The human 5-HT3R-As cDNA contains an open reading frame of 1434 base pairs predicting a 23-residue signal peptide and a mature receptor subunit of 455 amino acids. The human subunit exhibits 85% and 84% amino acid sequence identity to the mouse and rat receptors, respectively. Anti-5-HT3R polyclonal antibody shows immunoreactivity in the forebrain (olfactory regions, hippocampal formation and amygdala), sensory and motor nuclei of brainstem and spinal cord. 5-Ht3a is a member of the 5-HT3 receptor subfamily of ligand gated ion channels, as opposed to all other known 5-HT receptor subtypes that are G protein-coupled receptors.

HTR3A antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HTR3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR