Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPD1 Rabbit Polyclonal Antibody

Supplier: Prosci
49-288
HSP60, a 60 kDa member of Chaperonin family, is a mitochondrial protein that participates in protein folding, transport and secretion. HSP60 is a multi-functional protein whose roles include accelerating the maturation of pro-caspase 3 during apoptosis, LPS-binding, acting as a ligand for innate immune system and also binding to the surface of innate immune cells and stimulating proinflammatory reactivity. It induces the expression of IL-12 and IL-15 and also induces release of TNF-alpha, NO and IL-6. Reports suggest interaction of Hsp60 with integrin molecules during metastasis. Mutations in Hsp60 gene cause autosomal recessive spastic paraplegia 13. Hsp60 has been found to be involved in development of auto-immune disorders such as Rheumatoid arthritis, insulin-dependent diabetes mellitus, systemic sclerosis and atherosclerosis.

HSPD1 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 2.5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HSPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chimpanzee, Chicken, Dog, Human, Monkey




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
50 µg 49-288 10076-280 Each (50µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol HSPD1
Antigen Name Heat Shock 60kDa Protein 1 (Chaperonin)
Reactivity Chicken, Bovine, Monkey, Chimpanzee, Dog, Human
Conjugation Unconjugated
Clonality Polyclonal
Host Rabbit
Western Blot Yes
ImmunoChemistry Yes
Antigen Synonyms GROEL, Chaperonin 60, HSP60, HSP65, HSP-60, SPG13, CPN60, HLD4, HuCHA60, Mitochondrial Matrix Protein P1, Heat Shock Protein 65
Immunogen HSPD1 antibody was raised against amino acids 350 - 400 of HSPD1 (Human).
Form Liquid

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty