Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPA5 Rabbit Polyclonal Antibody

Supplier: Prosci
49-287
10076-278
Anti-HSPA5 Rabbit Polyclonal Antibody
Grp78, is a 78 kDa glucose-regulated protein, also known as BiP or immunoglobulin heavy chain binding protein, is a stress-response protein which is induced by agents or conditions that adversely affect endoplasmic reticulum (ER) function. This protein is essential for the proper glycosylation, folding and assembly of many membrane bound and secreted proteins. Grp78 is critical for maintenance of cell homeostasis and the prevention of apoptosis. Grp78 protein levels have been shown to be a reliable biomarker of hypoglycermia as well as serving a neuroprotective function in neurons exposed to glutamate and oxidative stress. Grp78 levels are reduced in the brains of Alzheimer's Disease patients and decreased expression of Grp78 is found associated with missense mutations in the human presenilin-1 (PS1) gene. The induction of the Grp78 protein has been associated with the development of drug-resistance to antitumor drugs.

HSPA5 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 μg/mL, and immunofluorescence starting at 10 μg/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: HSPA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty