Anti-HIST1H4A Rabbit Polyclonal Antibody

Supplier: ProSci
49-285
10078-786EA 684.46 USD
10078-786
Anti-HIST1H4A Rabbit Polyclonal Antibody
Antibodies
Histones are relatively small proteins that contain a high proportion of positively charged amino acids, e.g., arginine and lysine. This positive charge allows histones to bind tightly to the negatively charged DNA. There are five basic types of histone molecules called H1, H2A, H2B, H3, and H4. Histones are abundantly present in virtually all eukaryotic cells.

Histone H4 antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS, 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HIST1H4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Drosophila, Xenopus, Human, Monkey, Mouse, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR