Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-BMP10 Rabbit Polyclonal Antibody

Supplier: Prosci

49-267
10078-750EA 467.84 USD InStock
10078-750
Anti-BMP10 Rabbit Polyclonal Antibody
Antibodies
Bone Morphogenic Protein 10 is a member of the transforming growth factor-beta superfamily. In mouse heart, BMP-10 expression is localized to the trabeculated part of the common ventricular chamber and to the bulbus cordis region, the atrial wall and in the adult right atrium. BMP-10 has recently been implicated in postnatal cardiac hypertrophy.

BMP10 antibody can be used in ELISA starting at 1:10000 - 1:40000, Western Blot starting at 1:500 - 1:3000, and immunohistochemistry starting at 10 - 20 μg/mL.

Protein G Column

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: BMP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty