Anti-MCM7 Rabbit Polyclonal Antibody

49-256
10078-730EA 491.23 USD
10078-730
Anti-MCM7 Rabbit Polyclonal Antibody
CDC47/MCM7 participates in the regulation of mammalian DNA replication allowing DNA to replicate only once each cycle. It exists in two forms. The levels of the bound form in the nucleus have been shown to gradually decrease during S phase progression. Stimulation of cells to enter the cell cycle results in induction of CDC47/MCM7 expression.

MCM7 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: MCM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR