Anti-LAT Mouse Monoclonal Antibody [clone: [LAT1111]]

Supplier: Prosci

49-251
10078-722EA 561.89 USD
10078-722
Anti-LAT Mouse Monoclonal Antibody [clone: [LAT1111]]
Antibodies
LAT antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide.

Store LAT antibody at 4 °C.

Type: Primary
Antigen: LAT
Clonality: Monoclonal
Clone: [LAT1111]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR