Anti-PRMT7 Rabbit Polyclonal Antibody

Supplier: Prosci

49-229
10074-484EA 582.74 USD
10074-484
Anti-PRMT7 Rabbit Polyclonal Antibody
Antibodies
Protein arginine methylation is an important posttranslational modification in eukaryotic cells, which has been implicated in RNA processing and trafficking, receptor-mediated signaling, and transcription. The PRMT proteins are recruited by C-terminal activation domain-2 of the SRC (steroid receptor coactivator) GRIP-1, whereas p300 interacts with activation domain-1 in the C terminus. have identified a mammalian arginine N-methyltransferase, PRMT7, that can catalyze the formation of omega-NG-monomethylarginine in peptides. In contrast to the other members of the PRMT family, PRMT7 was not able to methylate GST-fibrillarin or myelin basic protein or histone H2A, in vitro. It is also structurally different from other PRMTS in containing two putative AdoMet-binding motifs. Both of these domains are required for enzymatic activity.

PRMT7 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PRMT7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR