Anti-PAPD7 Rabbit Polyclonal Antibody

Supplier: ProSci
49-227
10078-712EA 791.45 USD
10078-712
Anti-PAPD7 Rabbit Polyclonal Antibody
Antibodies
TRF4-1 is a nuclear protein whose expression is cell cycle-regulated at a post-transcriptional level. Two genes, TRF4-1 and TRF4-2, belonging to this family have been identified. TRF4 is also required to maintain a different aspect of higher order chromosome structure, sister chromatid cohesion.

POLS antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS containing 0.2% gelatin and 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PAPD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Gibbon, Gorilla, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR