Anti-MSH2 Rabbit Polyclonal Antibody

Supplier: Prosci

49-219
10078-696EA 582.9 USD
10078-696
Anti-MSH2 Rabbit Polyclonal Antibody
Antibodies
Homologous to the E. coli gene MutS, mutation of this gene is associated with hereditary nonpolyposis colorectal cancer (HNPCC).

MSH2 antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MSH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR