Anti-CD9 Mouse Monoclonal Antibody [clone: [HI9a]]

Supplier: ProSci
49-210
10078-682EA 674.53 USD
10078-682
Anti-CD9 Mouse Monoclonal Antibody [clone: [HI9a]]
Antibodies
CD9 is a member of the tetraspan family of membrane proteins. First described on non-T acute lymphoblastic leukemia cells and developing B-lymphocytes it has since been shown to be involved in platelet activation and aggregation, gamete fusion, cell adhesion, cell motility, tumor metastasis, and intracellular signaling in the nervous system.

CD9 antibody can be used in immunohistochemistry starting at 10 ?g/mL.

Affinity Chromatography

PBS, pH 7.2, 0.09% sodium azide.

Store CD9 antibody undiluted at 4 °C.

Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: [HI9a]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Bovine, Dog, Horse, Human, Rabbit, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR